1-Methyl-6,7-dimethoxy-3,4-dihydroisoquinoline 99+%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6,7-Dimethoxy-1-methyl-3,4-dihydroisoquinoline

265970010 265970050
ACRO265970010EA 83.7 EUR
ACRO265970010 ACRO265970050
1-Methyl-6,7-dimethoxy-3,4-dihydroisoquinoline 99+%
1-Methyl-6,7-dimethoxy-3,4-dihydroisoquinoline

Appearance: Light yellow to beige Crystalline powder

Formel: C₁₂H₁₅NO₂
Molecular Weight: 205,257 g/mol
Schmelzpunkt: 103…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040276
CAS-Nummer: 4721-98-6

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 103 to 107 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR