α-Hydroxy-γ-butyrolacton 99%
Lieferant: Thermo Scientific
Achtung
Synonyme: 3-Hydroxytetrahydrofuran-2-one, Dihydro-3-hydroxy-2(3H)-furanone, 3-Hydroxydihydrofuran-2(3H)-one
275070250 275070050
ACRO275070250EA 192 EUR
ACRO275070250 ACRO275070050
α-Hydroxy-γ-butyrolacton 99%
alpha-Hydroxy-gamma-butyrolactone
Appearance: Clear colorless to yellow Viscous liquid
Formel: C₄H₆O₃
Molecular Weight: 102,09 g/mol
Siedepunkt: 85…90 °C (,8mbar)
Dichte: 1,313 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00134268
CAS-Nummer: 19444-84-9
Bestellen
Spezifikation Testergebnisse
Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
GC ≥98.5 %