L-Allylglycine 98%, 98% ee

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (S)-(-)-alpha-Allylglycine, H-Ally-Gly-OH, H-Gly(Ally)-OH, 2-Allyl-L-glycine, (S)-(-)-2-Amino-4-pentenoic acid, (2S)-2-Aminopent-4-enoic acid, L-2-Amino-4-pentenoic acid L-Allylglycine L-alpha-Allylglycine, (S)-2-Allylglycine

275412500 275410010
ACRO275412500EA 93.1 EUR
ACRO275412500 ACRO275410010
L-Allylglycine 98%, 98% ee
L-Allylglycine

CAS No.: 16338-48-0

Formel: C₅H₉NO₂
Molecular Weight: 115.132 g/mol
Schmelzpunkt: 253…255 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00002627
CAS-Nummer: 16338-48-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
TLC ≥97.5 %
Enantiomeric excess ≥97.5 %
Specific optical rotation -35° to -39° (20°C, 589 nm) (c=4, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR