N-(tert-Butoxycarbonyl)-L-alanin 99+%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Alpha-t-Boc-L-alanine, (S)-2-((tert-Butoxycarbonyl)amino)propanoic acid, BOC-L-Alanine, Boc-Ala-OH

275490100 275490500
ACRO275490100EA 34 EUR
ACRO275490100 ACRO275490500
N-(tert-Butoxycarbonyl)-L-alanin 99+%
N-Boc-L-alanine

CAS No.: 15761-38-3

Formel: C₈H₁₅NO₄
Molecular Weight: 189.21 g/mol
Schmelzpunkt: 81…84 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00037225
CAS-Nummer: 15761-38-3

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Melting point 81 to 85 °C
HPLC ≥99.0 % (ELSD)
Specific optical rotation -25° to -27° (20 °C, 589 nm) (c=2, CH3COOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR