N-[(Benzyloxy)carbonyl]-L-valin 99+%

Lieferant: Thermo Scientific

Synonyme: N-Cbz-L-valine, Z-L-Val-OH, Z-Val-OH

275940050 275940250
ACRO275940050EA 29 EUR
ACRO275940050 ACRO275940250
N-[(Benzyloxy)carbonyl]-L-valin 99+%
N-Carbobenzyloxy-L-valine

Appearance: White to off-white Crystalline powder

Formel: C₁₃H₁₇NO₄
Molecular Weight: 251,3 g/mol
Schmelzpunkt: 58…60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008922
CAS-Nummer: 1149-26-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.2 %
HPLC ≥98.0 %
Specific optical rotation -3.2° to -4.8° (20°C, 589 nm) (c=2, acetic acid)
Titration with NaOH ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR