cis-2-Amino-1-cyclopentanecarboxylic acid 98+%
Lieferant: Thermo Fisher Scientific
277745000
277740020
ACRO277745000EA
106
EUR
ACRO277745000
ACRO277740020
cis-2-Amino-1-cyclopentanecarboxylic acid 98+%
cis-2-Amino-1-cyclopentanecarboxylic acid
CAS No.: 37910-65-9
Formel:
C₆H₁₁NO₂ Molecular Weight: 129.159 g/mol Schmelzpunkt: 218…220 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143982 CAS-Nummer: 37910-65-9 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | 98.0 to 102.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...