(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptan-2,3-diol 98%

Lieferant: Thermo Scientific

Synonyme: (1S,2S,3R,5S)-(+)-2,3-Pinanediol, (1S,2S,3R,5S)-(+)-Pinanediol

279550025 279550100
ACRO279550025EA 44.9 EUR
ACRO279550025 ACRO279550100
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptan-2,3-diol 98%
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol

CAS No.: 18680-27-8

Formel: C₁₀H₁₈O₂
Molecular Weight: 170,25 g/mol
Schmelzpunkt: 52…60 °C
Flammpunkt: >110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00077851
CAS-Nummer: 18680-27-8

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Crystalline powder or crystals or mass
Infrared spectrum Conforms
Melting point 52 to 60 °C
GC ≥97.5 %
Specific optical rotation +7° to +10° (20 °C, 589 nm) (c=6.5, toluene)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR