(2S,3S)-2-Amino-3-methoxybutanoic acid 98%

Lieferant: Thermo Scientific

290032500 290030010
ACRO290032500EA 192 EUR
ACRO290032500 ACRO290030010
(2S,3S)-2-Amino-3-methoxybutanoic acid 98%
(2S,3S)-2-Amino-3-methoxybutanoic acid

CAS No.: 104195-80-4

Formel: C₅H₁₁NO₃
Schmelzpunkt: 220…224 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00142985
CAS-Nummer: 104195-80-4

Bestellen

Spezifikation Testergebnisse

Appearance White Powder or flakes
Infrared spectrum Conforms
Specific optical rotation +25° to +29° (25°C, 589 nm) (C=0.5, 6 N HCl)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR