N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) 96%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-(Chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane Bis(tetrafluoroborate), F-TEDA-BF4, 1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate)
291490050
291490250
291491000
ACRO291490050EA
37
EUR
ACRO291490050
ACRO291490250
ACRO291491000
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) 96%
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)
CAS No.: 140681-55-6
Formel:
C₇H₁₄B₂ClF₉N₂ Molecular Weight: 354.26 g/mol Schmelzpunkt: 260 °C (Zers.) Flammpunkt: >54 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00142607 CAS-Nummer: 140681-55-6 UN: 3088 ADR: 4.2 II |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
Titration Iodometric | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...