3-Phenyl-1H-pyrazol 99+%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3(5)-Phenyl-1H-pyrazole, 3-Phenylpyrazole
296030025
296030100
ACRO296030025EA
45.5
EUR
ACRO296030025
ACRO296030100
3-Phenyl-1H-pyrazol 99+%
3-Phenyl-1H-pyrazole
Formel:
C₉H₈N₂ Molecular Weight: 144,18 g/mol Schmelzpunkt: 75…77 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00159654 CAS-Nummer: 2458-26-6 |
Spezifikation Testergebnisse
Appearance | White to yellow to light orange Powder |
Infrared spectrum | Conforms |
Melting point | 74 to 80 °C |
GC | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...