(R)-Glycidyl-3-nitrobenzenesulphonate 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (R)-(-)-Glycidyl nosylate, 3-Nitrobenzenesulphonic acid (R)-Glycidyl Ester, (R)-Oxiran-2-ylmethyl 3-nitrobenzenesulphonate

296410025 296410100
ACRO296410025EA 57.5 EUR
ACRO296410025 ACRO296410100
(R)-Glycidyl-3-nitrobenzenesulphonate 98%
(R)-Glycidyl-3-nitrobenzenesulphonate

CAS No.: 115314-17-5

Formel: C₉H₉NO₆S
Molecular Weight: 259,23 g/mol
Schmelzpunkt: 59…63 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00013436
CAS-Nummer: 115314-17-5

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow Crystalline powder or needles
Infrared spectrum Conforms
Melting point 59 to 63 °C
GC ≥97.5 %
Specific optical rotation (20 °C, 589 nm) (c=2, CHCl3) < -21.5°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR