4-Methylpent-2-enal 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Methyl-2-pentenal
298350250
298351000
ACRO298350250EA
120
EUR
ACRO298350250
ACRO298351000
4-Methylpent-2-enal 95%
4-Methylpent-2-enal
Appearance: Clear colorless to light yellow Liquid
Formel:
C₆H₁₀O Siedepunkt: 70 °C (75 mmHg) Dichte: 0,844 g/cm³ Flammpunkt: 32 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00800501 CAS-Nummer: 5362-56-1 UN: 1993 ADR: 3,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4360 to 1.4440 (20°C, 589 nm) |
GC | ≥94 % |
Specific gravity | (25°C) 0.831 to 0.841 |
Acid value | ≤15 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...