(S)-2-Amino-3-hydroxy-3-methylbutanoic acid 98%, 94% ee

Lieferant: Thermo Scientific

Synonyme: (S)-(+)-2-Amino-3-hydroxy-3-methylbutanoic acid, H-Val(3-OH)-OH

299142500 299140010
ACRO299142500EA 131 EUR
ACRO299142500 ACRO299140010
(S)-2-Amino-3-hydroxy-3-methylbutanoic acid 98%, 94% ee
(S)-2-Amino-3-hydroxy-3-methylbutanoic acid

CAS No.: 2280-27-5

Formel: C₅H₁₁NO₃
Molecular Weight: 133,15
Lagertemperatur: Raumtemperatur
MDL: MFCD00800535
CAS-Nummer: 2280-27-5

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
TLC ≥97.5 %
Enantiomeric excess ≥93 % (HPLC)
Specific optical rotation +11° to +14° (20°C, 589 nm) (c=2, 6 N HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR