2-(3-Methoxyallylidene)malonic acid dimethyl ester 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-(3-Methoxyallylidene)malonic acid dimethyl ester
302090025
302090100
ACRO302090025EA
75.2
EUR
ACRO302090025
ACRO302090100
2-(3-Methoxyallylidene)malonic acid dimethyl ester 95%
2-(3-Methoxyallylidene)malonic acid dimethyl ester
CAS No.: 41530-32-9
Formel:
C₉H₁₂O₅ Molecular Weight: 200,19 g/mol Siedepunkt: 120…125 °C (,3 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00800589 CAS-Nummer: 41530-32-9 |
Spezifikation Testergebnisse
Appearance | Clear yellow-green Low melting solid |
Infrared spectrum | Conforms |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...