(S)-Methyl-2-aminopropanoate hydrochloride 99%

Lieferant: Thermo Scientific

Synonyme: H-Ala-OMe·HCl, H-Ala-OMe.HCl, L-Alanine methyl ester hydrochloride, (S)-2-Aminopropionic Acid Methyl Ester Hydrochloride

308360250 308360050
ACRO308360250EA 102 EUR
ACRO308360250 ACRO308360050
(S)-Methyl-2-aminopropanoate hydrochloride 99%
(S)-Methyl-2-aminopropanoate hydrochloride

Appearance: White to off-white Powder

Formel: C₄H₉NO₂·HCl
Molecular Weight: 139,6 g/mol
Schmelzpunkt: 109…111 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00063663
CAS-Nummer: 2491-20-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Specific optical rotation +6° to +8° (24°C, 589 nm) (c=1.6, CH3OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR