Buthionine sulphoximine 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
DL-Buthionine (S,R)-sulphoximine
309475000
309470050
ACRO309475000EA
130
EUR
ACRO309475000
ACRO309470050
Buthionine sulphoximine 99%
Buthionine sulphoximine
CAS No.: 5072-26-4
Formel:
C₈H₁₈N₂O₃S Molecular Weight: 222,303 g/mol Schmelzpunkt: 215 °C (Zersetzung) Lagertemperatur: Kühlschrank |
MDL:
MFCD00070309 CAS-Nummer: 5072-26-4 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...