L-Aspartyl-L-phenylalanine
Lieferant: Thermo Fisher Scientific
310021000
310025000
ACRO310021000EA
86.5
EUR
ACRO310021000
ACRO310025000
L-Aspartyl-L-phenylalanine
Proteine und Peptide
CAS No.: 13433-09-5
- Infrared spectrum: Conforms
- Appearance of solution: (5% in CH₃COOH 80%) clear colorless
- Specific optical rotation: 10.0 to 13.0° (24 °C, 589 nm) (c=1, 0.5 N HCl)
- Melting point: 236 to 239 °C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...