Sodium-tert-pentyloxide 95%, rein
Lieferant: Thermo Scientific
Gefahr
Synonyme: Sodium tert-pentoxide, Sodium tert-pentoxide, 95%, tert-Amyl alcohol sodium salt
327551000 327555000
ACRO327551000EA 61.4 EUR
ACRO327551000 ACRO327555000
Sodium-tert-pentyloxide 95%, rein
Sodium-tert-pentyloxide
Appearance: Cream to tan Powder
Formel: C₅H₁₁NaO
Molecular Weight: 110,132 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00064245
CAS-Nummer: 14593-46-5
UN: 3206
ADR: 4.2,II
Bestellen
Spezifikation Testergebnisse
Appearance Cream to tan Powder
Titration with HClO4 ≥94 %