Nifedipin 98+%

Lieferant: Thermo Scientific
Achtung

Synonyme: Dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Adalat, 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic Acid Dimethyl Ester

329270050 329270010 329270250
ACRO329270050EA 102 EUR
ACRO329270050 ACRO329270010 ACRO329270250
Nifedipin 98+%
Nifedipin

CAS No.: 21829-25-4

Formel: C₁₇H₁₈N₂O₆
Molecular Weight: 346,34 g/mol
Schmelzpunkt: 171…175 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00057326
CAS-Nummer: 21829-25-4

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Powder
Infrared spectrum Conforms
Melting point 171 to 175 °C
Loss on drying ≤0.5 %
HPLC ≥98.0 %
Residue after ignition ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR