Tamoxifen citrate 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(Z)-1-(4-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate
329320010
329320050
ACRO329320010EA
247
EUR
ACRO329320010
ACRO329320050
Tamoxifen citrate 98%
Tamoxifen citrate
CAS No.: 54965-24-1
Formel:
C₂₆H₂₉NO Molecular Weight: 563,65 g/mol Schmelzpunkt: 140…144 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00058321 CAS-Nummer: 54965-24-1 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...