6,7-Dihydro-4(5H)-benzofuranone 98%
Lieferant: Thermo Scientific
Synonyme:
6,7-Dihydrobenzofuran-4(5H)-one
330080050
330080250
ACRO330080050EA
98.8
EUR
ACRO330080050
ACRO330080250
6,7-Dihydro-4(5H)-benzofuranone 98%
6,7-Dihydro-4(5H)-benzofuranone
Appearance: Clear yellow to brown Liquid or low melting solid
Formel:
C₈H₈O₂ Molecular Weight: 136,15 g/mol Siedepunkt: 115…118 °C (16 mmHg) Schmelzpunkt: 30…34 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00215511 CAS-Nummer: 16806-93-2 |
Spezifikation Testergebnisse
Appearance | Clear yellow to brown Liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...