3-(2-Thienyl)propanoic acid 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3-(2-Thienyl)propionic Acid
330110010
330110050
ACRO330110010EA
37
EUR
ACRO330110010
ACRO330110050
3-(2-Thienyl)propanoic acid 98%
3-(2-Thienyl)propanoic acid
Appearance: White to beige Crystals or needles
Formel:
C₇H₈O₂S Molecular Weight: 156,199 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047093 CAS-Nummer: 5928-51-8 UN: 3261 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | White to beige Crystals or needles |
Infrared spectrum | Conforms |
Melting point | 45 to 49 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...