2-Methylallylamine 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3-Amino-2-methyl-1-propene
330700010
330700050
ACRO330700010EA
87.3
EUR
ACRO330700010
ACRO330700050
2-Methylallylamine 97%
2-Methylallylamine
Appearance: Clear colorless to light yellow Liquid
Formel:
C₄H₉N Molecular Weight: 71,123 g/mol Siedepunkt: 78 °C (760 mmHg) Dichte: 0,78 g/cm³ Flammpunkt: –28 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00053646 CAS-Nummer: 2878-14-0 UN: 1993 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4290 to 1.4320 (20°C, 589 nm) |
Titration with HClO4 | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...