(5R)-5-(Hydroxymethyl)-2-pyrrolidinon 98%
Lieferant: Thermo Scientific
|
Achtung
|
333560010
333560050
333560250
ACRO333560010EA
25.1
EUR
ACRO333560010
ACRO333560050
ACRO333560250
(5R)-5-(Hydroxymethyl)-2-pyrrolidinon 98%
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone
CAS No.: 66673-40-3
Formel:
C₅H₉NO₂ Molecular Weight: 115,13 g/mol Siedepunkt: 147…149 °C Schmelzpunkt: 77…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077791 CAS-Nummer: 66673-40-3 |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder or flakes |
Infrared spectrum | Conforms |
Melting point | 77 to 84 °C |
HPLC | ≥97.5 % |
Specific optical rotation | -27° to -35° (20 °C, 589 nm) (c=1, C2H5OH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...