4-Methylumbelliferyl-β-D-galactopyranoside

Lieferant: Thermo Scientific

Synonyme: 4-Methylumbelliferyl β-D-galactopyranoside, 4-Methyl-7-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one

337210010 337210050
ACRO337210010EA 75 EUR
ACRO337210010 ACRO337210050
4-Methylumbelliferyl-β-D-galactopyranoside
4-Methylumbelliferyl-β-D-galactopyranoside

CAS No.: 6160-78-7

Formel: C₁₆H₁₈O₈
Molecular Weight: 338,312 g/mol
Schmelzpunkt: 248 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00036773
CAS-Nummer: 6160-78-7

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
TLC Complies
Appearance of solution (1% in pyridine) clear colorless
Specific optical rotation -62° to -66° (20°C, 589 nm) (c=1, pyridine, after 24h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR