4-Amino-6-methoxypyrimidine 97%
Lieferant: Thermo Scientific
|
Achtung
|
337450050
337450250
ACRO337450050EA
67.2
EUR
ACRO337450050
ACRO337450250
4-Amino-6-methoxypyrimidine 97%
4-Amino-6-methoxypyrimidine
Appearance: White to yellow Crystalline powder
Formel:
C₅H₇N₃O Molecular Weight: 125,1286 g/mol Schmelzpunkt: 154…159 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00129983 CAS-Nummer: 696-45-7 |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 154 to 159 °C |
Titration with HClO4 | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...