Triallyl isocyanurate 98% stabilisiert

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1,3,5-Triallyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, Isocyanuric Acid Triallyl Ester, 1,3,5-Triallyl-1,3,5-triazinane-2,4,6-trione

342121000 342125000
ACRO342121000EA 42.4 EUR
ACRO342121000 ACRO342125000
Triallyl isocyanurate 98% stabilisiert
Triallyl isocyanurate

Appearance: Clear colorless to yellow Viscous liquid or solid

Formel: C₁₂H₁₅N₃O₃
Molecular Weight: 249.27 g/mol
Siedepunkt: 149…152 °C (4 mmHg)
Schmelzpunkt: 24.8 °C
Dichte: 1.159 g/cm³
Flammpunkt: 245 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006554
CAS-Nummer: 1025-15-6

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Viscous liquid or solid
Infrared spectrum Conforms
Refractive index 1.5120 to 1.5140 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 100 to 1200 ppm MEHQ

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR