Captopril 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(S)-(-)-1-(3-Mercapto-2-methyl-1-oxopropyl)-L-proline, (S)-(-)-1-(3-Mercapto-2-methylpropionyl)-L-proline, (S)-1-((S)-3-mercapto-2-methylpropanoyl)pyrrolidine-2-carboxylic acid
343700010
343700050
343700250
ACRO343700010EA
82.6
EUR
ACRO343700010
ACRO343700050
ACRO343700250
Captopril 98%
Captopril
Appearance: White Crystalline powder
Formel:
C₉H₁₅NO₃S Molecular Weight: 217,28 g/mol Schmelzpunkt: 103…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00168073 CAS-Nummer: 62571-86-2 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 103 to 108 °C |
HPLC | ≥97.5 % |
Specific optical rotation | -125° to -134° (20 °C, 589 nm) (c=1, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...