tert-Butyl (3S)-3-amino-4-phenylbutanoate 97%
Lieferant: Thermo Scientific
|
Gefahr
|
344461000
344465000
ACRO344461000EA
97.8
EUR
ACRO344461000
ACRO344465000
tert-Butyl (3S)-3-amino-4-phenylbutanoate 97%
tert-Butyl (3S)-3-amino-4-phenylbutanoate
Appearance: White to light yellow Low melting solid
Formel:
C₁₄H₂₁NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798308 CAS-Nummer: 120686-17-1 UN: 3259 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | White to light yellow Low melting solid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...