BAPTA tetrasodium salt hydrate 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
BAPTA-Na4 hydrate, 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid tetrasodium Salt hydrate, Tetrasodium-1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate hydrate
401650010
401650050
ACRO401650010EA
113
EUR
ACRO401650010
ACRO401650050
BAPTA tetrasodium salt hydrate 95%
BAPTA tetrasodium salt hydrate
CAS No.: 126824-24-6
Formel:
C₂₂H₂₀N₂Na₄O₁₀·xH₂O Molecular Weight: 564.37 g/mol (anhy) Lagertemperatur: Gefrierschrank |
MDL:
MFCD02683087 CAS-Nummer: 336624-09-0 |
Spezifikation Testergebnisse
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
Titration Complexometric | ≥94.0 % |
Water | ≤12 % (K.F.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...