5(6)-Carboxyfluorescein
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5(6)-FAM, 3′,6′-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthene]-ar-carboxylic acid
404100050
404105000
ACRO404100050EA
127
EUR
ACRO404100050
ACRO404105000
5(6)-Carboxyfluorescein
5(6)-Carboxyfluorescein
Appearance: Orange to brown Crystalline powder and/or chunks
Formel:
C₄₂H₂₄O₁₄ Molecular Weight: 752.63 g/mol Schmelzpunkt: 275 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151081 CAS-Nummer: 72088-94-9 |
Spezifikation Testergebnisse
Appearance | Orange to brown Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Molar absorption coefficient | ≥72000 l/(mol.cm) (at Lamda max 492 nm, in 0.01N NaOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...