Cumarin 343, rein Laserqualität

Lieferant: Thermo Scientific

ACRO405681000EA 70.8 EUR
ACRO405681000 ACRO405685000
Cumarin 343, rein Laserqualität
Coumarin 343

Appearance: Orange Crystalline powder

Formel: C₁₆H₁₅NO₄
Molecular Weight: 285,30 g/mol
Schmelzpunkt: 240 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051335
CAS-Nummer: 55804-65-4

Bestellen

Spezifikation Testergebnisse

Appearance Orange Crystalline powder
Infrared spectrum Conforms
UV Lambda max 409 nm (in Basic Ethanol)
Fluorescence max 470 nm (in Basic Ethanol)
Optimum lasing 518 nm (in Methanol)
Suitability (for use as a laser dye) Passes test
Molar absorption coefficient 19900 l/(mol.cm) (at Lambda max)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR