4,4'-Diaminostilbene-2,2'-disulphonic Acid 95%
Lieferant: Thermo Scientific
Synonyme:
Amsonic acid, 4,4-Diamino-2,2-stilbenedisulphonic acid
406411000
406415000
ACRO406411000EA
73.8
EUR
ACRO406411000
ACRO406415000
4,4'-Diaminostilbene-2,2'-disulphonic Acid 95%
4,4'-Diaminostilbene-2,2'-disulphonic Acid
Spezifikation Testergebnisse
Appearance | Cream to yellow-brown Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...