Diethyl iminodiacetate 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Iminodiacetic Acid Diethyl Ester, Diethyl 2,2'-azanediyldiacetate

407320250 407321000 407320050
ACRO407320250EA 124 EUR
ACRO407320250 ACRO407321000 ACRO407320050
Diethyl iminodiacetate 98%
Diethyl iminodiacetate

Appearance: Clear colorless to yellow Liquid

Formel: C₈H₁₅NO₄
Molecular Weight: 189,21 g/mol
Siedepunkt: 248 °C (760 mmHg)
Dichte: 1,08 g/cm³
Flammpunkt: 104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041925
CAS-Nummer: 6290-05-7

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4350 to 1.4380 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR