Ferrozine mono-sodium salt hydrate 98+%, rein

Lieferant: Thermo Scientific
Achtung

Synonyme: 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazin-4,4''-disulfonsäure Mononatriumsalz Hydrat

410570050 410570010 410570250
ACRO410570050EA 160 EUR
ACRO410570050 ACRO410570010 ACRO410570250
Ferrozine mono-sodium salt hydrate 98+%, rein
Ferrozine mono-sodium salt hydrate

FerroZine™ iron reagent

Formel: C₂₀H₁₃N₄NaO₆S₂·H₂O
Molecular Weight: 492,47 g/mol (anhy)
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00078417
CAS-Nummer: 1266615-85-3

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Water ≤8 % (K.F.)
Specific extinction E (1%/1cm) ≤0.015 (562 nm in water)
Titration with NaOH ≥98.0 % (On dry substance)
Suitability (for Cu + Fe determination) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR