(R)-4-Phenylbutan-2-amine 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(R)-(-)-1-Methyl-3-phenylpropylamine
414760010
414760050
ACRO414760010EA
152
EUR
ACRO414760010
ACRO414760050
(R)-4-Phenylbutan-2-amine 98%
(R)-4-Phenylbutan-2-amine
Appearance: Clear colorless Liquid
Formel:
C₁₀H₁₅N Molecular Weight: 149,237 g/mol Dichte: 0,936 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00145208 CAS-Nummer: 937-52-0 UN: 2922 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5130 to 1.5150 (20°C, 589 nm) |
Specific optical rotation | -10° to -12° (20°C, 589 nm) (neat) |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...