Nilrot 99%, rein

Lieferant: Thermo Scientific

ACRO415710010EA 842 EUR
ACRO415710010 ACRO415711000
Nilrot 99%, rein
Nilrot
Formel: C₂₀H₁₈N₂O₂
Molecular Weight: 318,38 g/mol
Schmelzpunkt: 203…205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011639
CAS-Nummer: 7385-67-3

Bestellen

Spezifikation Testergebnisse

Appearance Dark green to dark red to brown Powder
Infrared spectrum Conforms
UV spectrum Complies
UV spectrophotometry ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR