Phenolphthalein disulphate tripotassium salt trihydrate 98%, rein

Lieferant: Thermo Scientific

417220050 417220100
ACRO417220050EA 83.4 EUR
ACRO417220050 ACRO417220100
Phenolphthalein disulphate tripotassium salt trihydrate 98%, rein
Phenolphthalein disulphate tripotassium salt trihydrate

Appearance: White to off-white Crystalline powder

Formel: C₂₀H₁₃K₃O₁₁S₂.3H₂O
Schmelzpunkt: 230 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00150195
CAS-Nummer: 1080664-81-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR