TCSA 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,3',4',5-Tetrachlorosalicylanilide, 3,5-Dichloro-N-(3,4-dichlorophenyl)-2-hydroxybenzamide

420230050 420230250
ACRO420230050EA 49.2 EUR
ACRO420230050 ACRO420230250
TCSA 99%
TCSA

Appearance: Off-white to beige Crystalline powder

Formel: C₁₃H₇Cl₄NO₂
Molecular Weight: 351.004 g/mol
Schmelzpunkt: 162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041745
CAS-Nummer: 1154-59-2
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
UV spectrophotometry ≥98.5 %
HPLC ≥94 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR