Ethyl-5-nitroindole-2-carboxylate 95%

Lieferant: Thermo Scientific

Synonyme: Ethyl 5-nitro-1H-indole-2-carboxylate

424660025 424660100
ACRO424660025EA 91.5 EUR
ACRO424660025 ACRO424660100
Ethyl-5-nitroindole-2-carboxylate 95%
Ethyl-5-nitroindole-2-carboxylate

CAS No.: 16732-57-3

Formel: C₁₁H₁₀N₂O₄
Molecular Weight: 234,211 g/mol
Schmelzpunkt: 220…225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216477
CAS-Nummer: 16732-57-3

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to brown Powder
Infrared spectrum Conforms
Melting point 220 to 225 °C
Titration with TBAH ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR