3-Acetyl-2,4-dimethylpyrrole 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
102510100
102510500
ACRO102510100EA
148
EUR
ACRO102510100
ACRO102510500
3-Acetyl-2,4-dimethylpyrrole 98%
3-Acetyl-2,4-dimethylpyrrole
CAS No.: 2386-25-6
Formel:
C₈H₁₁NO Molecular Weight: 137.18 g/mol Schmelzpunkt: 136…139 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005221 CAS-Nummer: 2386-25-6 |
Spezifikation Testergebnisse
Appearance | Light yellow to pink to brown to purple Powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...