6-Aminopenicillansäure 96%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 6-APA 6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 6-amino-2,2-dimethyl-5-oxoperhydroazeto[2,1-b][1,3]thiazole-3-carboxylic acid, (2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, (+)-6-Aminopenicillanic acid, (2S,5R,6R)-(+)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid

104250250 104251000 104250100
ACRO104250250EA 83.6 EUR
ACRO104250250 ACRO104251000 ACRO104250100
6-Aminopenicillansäure 96%
6-Aminopenicillanic acid

CAS No.: 551-16-6

Formel: C₈H₁₂N₂O₃S
Molecular Weight: 216,26 g/mol
Schmelzpunkt: 197…203 °C (Zersetzung)
Lagertemperatur: Kühlschrank
MDL: MFCD00005176
CAS-Nummer: 551-16-6

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 197 to 203 °C
HPLC ≥95 %
pH 3.8 to 4.3 (4 % slurry in water)
Loss on heating ≤1.5 % (105 °C, 2 h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR