Ethyl-(triphenylphosphoranyliden)acetat 98+%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Ethyl triphenylphosphoranylidene-acetate, (Triphenylphosphoranylidene)acetic acid ethyl ester, (Carbethoxymethylene)triphenylphosphorane, Ethoxycarbonylmethylene-triphenylphosphorane, Ethyl 2-(triphenylphosphoranylidene)acetate

108290250 108291000 108295000
ACRO108290250EA 76.6 EUR
ACRO108290250 ACRO108291000 ACRO108295000
Ethyl-(triphenylphosphoranyliden)acetat 98+%
Ethyl-2-(triphenylphosphoranylidene)acetate

CAS No.: 1099-45-2

Formel: C₂₂H₂₁O₂P
Molecular Weight: 348.38 g/mol
Schmelzpunkt: 124…129 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00009183
CAS-Nummer: 1099-45-2

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 124 to 129 °C
HPLC ≥98.0 %
UV at 250 nm A: ≤0.05
Titration after Ion exchange ≥97.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR