3,3'-Diaminobenzidin Tetrahydrochlorid Hydrat, (max. 10% H₂O) 97%
Lieferant: Thermo Scientific
Gefahr
Synonyme: DAB HCl hydrate, 3,3',4,4'-Biphenyltetramine tetrahydrochloride hydrate, DAB tetrahydrochloride hydrate
112090050 112090250 112090010
ACRO112090050EA 68.5 EUR
ACRO112090050 ACRO112090250 ACRO112090010
3,3'-Diaminobenzidin Tetrahydrochlorid Hydrat, (max. 10% H₂O) 97%
3,3'-Diaminobenzidin Tetrahydrochlorid Hydrat
Formel: C₁₂H₁₄N₄·4HCl·H₂O
Molecular Weight: 360,11 g/mol (anhy)
Schmelzpunkt: 300 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD08273058
CAS-Nummer: 868272-85-9
Bestellen
Spezifikation Testergebnisse
Appearance Grey to brown-purple Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 % (on dry substance)
Loss on drying ≤10 % (120°C)