meso-Erythritol 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
meso-Erythritol, (2R,3S)-rel-Butane-1,2,3,4-tetraol, i-Erythritol
117820250
117821000
117825000
ACRO117820250EA
85.7
EUR
ACRO117820250
ACRO117821000
ACRO117825000
meso-Erythritol 99%
meso-Erythritol
CAS No.: 149-32-6
Formel:
C₄H₁₀O₄ Siedepunkt: 329…331 °C (760 mmHg) Schmelzpunkt: 117…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004710 CAS-Nummer: 149-32-6 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 118 to 122 °C |
Water | ≤1 % (K.F.) |
Solubility | (10 % in water) Clear colorless |
HPLC | ≥98.5 % (ELSD) |
Sodium (Na) | ≤0.005 % |
Heavy metals (as Pb) | ≤5 ppm |
Sulfur compounds (as S) | ≤0.002 % |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...