Ethyl-2-oxocyclopentaneacetate 95%

Lieferant: Thermo Scientific

Synonyme: 2-Oxocyclopentaneacetic Acid Ethyl Ester, Ethyl 2-oxocyclopentylacetate, Ethyl 2-(2-Oxocyclopentyl)acetate, 2-(2-Oxocyclopentyl)acetic Acid Ethyl Ester, Ethyl 2-(2-oxocyclopentyl)acetate

123040010 123040050
ACRO123040010EA 70.9 EUR
ACRO123040010 ACRO123040050
Ethyl-2-oxocyclopentaneacetate 95%
Ethyl-2-oxocyclopentaneacetate

CAS No.: 20826-94-2

Formel: C₉H₁₄O₃
Molecular Weight: 170,21 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00044715
CAS-Nummer: 20826-94-2

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94 %
Specific gravity 0.859 to 0.864
Specific optical rotation -15° to -21° (20°C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR