[(Diphenylmethylen)amino]acetonitril 99%

Lieferant: Thermo Scientific

Synonyme: N-(Diphenylmethylene)aminoacetonitrile

124140050 124140250
ACRO124140050EA 60.2 EUR
ACRO124140050 ACRO124140250
[(Diphenylmethylen)amino]acetonitril 99%
2-((Diphenylmethylene)amino)acetonitrile

CAS No.: 70591-20-7

Formel: C₁₅H₁₂N₂
Molecular Weight: 220,2700 g/mol
Schmelzpunkt: 83…86 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00009970
CAS-Nummer: 70591-20-7

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to light grey Powder or crystals
Infrared spectrum Conforms
Melting point 83 to 86 °C
HPLC ≥98.5 %
Identification A Passes test
Identification B Passes test
Specific optical rotation +12.0° to +12.8° (20 °C, 589 nm) (c=20, H2O) (on dried substance)
Titration with NaOH 99.5 to 101.0 % (on dried substance)
Sulfates ≤150 ppm
Chlorides ≤100 ppm
Calcium (Ca) ≤200 ppm
Impurity ≤350 ppm (as anhydrous oxalic acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR