Pamoic acid 98+%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4,4'-Methylenebis(3-hydroxy-2-naphthoic acid), Embonic acid

129721000 129725000
ACRO129721000EA 55.6 EUR
ACRO129721000 ACRO129725000
Pamoic acid 98+%
Pamoic acid

CAS No.: 130-85-8

Formel: C₂₃H₁₆O₆
Molecular Weight: 388,38 g/mol
Schmelzpunkt: >300 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004079
CAS-Nummer: 130-85-8

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to yellow-green Crystalline powder
Infrared spectrum Conforms
Titration excess NaOH 98.0 to 102.0 %
GC ≥97.0 %
Stabilizer ≤500 ppm BHT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR