2-Pyridinylessigsäurehydrochlorid 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(Pyridin-2-yl)acetic acid hydrochloride, 2-(Pyridin-2-yl)acetic acid HCl
131950050
131950250
ACRO131950050EA
54.5
EUR
ACRO131950050
ACRO131950250
2-Pyridinylessigsäurehydrochlorid 99%
2-Pyridylacetic acid hydrochloride
Appearance: White to beige Crystalline powder or crystals
Formel:
C₇H₇NO₂ Molecular Weight: 173.60 g/mol Schmelzpunkt: 135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012812 CAS-Nummer: 16179-97-8 |
Spezifikation Testergebnisse
Appearance | White to beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration Argentometric | ≥98.5 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...