2-(Bromacetyl)naphthalin 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
α-Bromo-2'-acetonaphthone, 2-Bromo-2'-acetonaphthone
147480100
147480500
ACRO147480100EA
62.7
EUR
ACRO147480100
ACRO147480500
2-(Bromacetyl)naphthalin 98%
2-(Bromacetyl)naphthalin
Formel:
C₁₂H₉BrO Molecular Weight: 249.11 g/mol Schmelzpunkt: 82…85 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00004109 CAS-Nummer: 613-54-7 UN: 3261 ADR: 8 II |
Spezifikation Testergebnisse
Appearance | Brown-purple Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 82 to 85 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...